Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alg6
Entrez Gene ID
320438
Gene Name
asparagine-linked glycosylation 6 (alpha-1,3,-glucosyltransferase)
Gene Aliases
E230028F23Rik
Chromosome Location
Chr.4: 99715599 - 99763460 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081264.1 NP_001074733.1 9 - 10 1016 96
GenBank mRNA AK157903.1 - 9 - 10 978 96
AK171887.1 - 9 - 10 969 96
BC050854.1 - 9 - 10 981 96
BC080867.1 - 9 - 10 956 96

View Full Product Details