Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc4a3
Entrez Gene ID
20536
Gene Name
solute carrier family 4 (anion exchanger), member 3
Gene Aliases
A930038D23Rik, Ae3
Chromosome Location
Chr.1: 75546041 - 75559431 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009208.3 NP_033234.2 16 - 17 2742 94
GenBank mRNA AK147218.1 - - 3079 94
BC066086.1 - 17 - 18 2616 94
BC080271.1 - 16 - 17 2737 94
M28383.1 - 16 - 17 2712 94

View Full Product Details