The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053272.2 | NP_444502.2 | 8 - 9 | 1533 | 63 |
GenBank mRNA | AK017937.1 | - | 8 - 9 | 1295 | 63 |
AK083737.1 | - | 8 - 9 | 1488 | 63 | |
AK129036.1 | - | 8 - 9 | 1533 | 63 | |
AY039762.1 | - | 8 - 9 | 1427 | 63 | |
BC019797.1 | - | 8 - 9 | 1477 | 63 |