Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gsg1
Entrez Gene ID
14840
Gene Name
germ cell-specific gene 1
Gene Aliases
-
Chromosome Location
Chr.6: 135237329 - 135254336 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080552.1 NP_001074021.1 3 - 4 542 130
NM_001080553.1 NP_001074022.1 3 - 4 695 130
NM_010352.2 NP_034482.2 4 - 5 755 130
GenBank mRNA AK006326.1 - 1 - 2 138 130
AK088285.1 - 1 - 2 138 130
BC023009.1 - 3 - 4 627 130
D87325.1 - 4 - 5 634 130

View Full Product Details