Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arhgap27
Entrez Gene ID
544817
Gene Name
Rho GTPase activating protein 27
Gene Aliases
2310069I04Rik, 5730442P18Rik, Gm21552, Sh3d20
Chromosome Location
Chr.11: 103331484 - 103363692 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205236.1 NP_001192165.1 13 - 14 2580 55
NM_133715.5 NP_598476.2 11 - 12 1549 55
GenBank mRNA AK046763.1 - 11 - 12 1550 55
AK143916.1 - 9 - 10 1021 55
AK154926.1 - 12 - 13 2871 55

View Full Product Details