The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205236.1 | NP_001192165.1 | 8 - 9 | 2062 | 88 |
NM_133715.5 | NP_598476.2 | 6 - 7 | 1031 | 88 | |
GenBank mRNA | AK046763.1 | - | 6 - 7 | 1032 | 88 |
AK143916.1 | - | 4 - 5 | 503 | 88 |