Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jakmip2
Entrez Gene ID
76217
Gene Name
janus kinase and microtubule interacting protein 2
Gene Aliases
6430702L21Rik, AI850334, D930046L20Rik
Chromosome Location
Chr.18: 43527096 - 43687773 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163637.1 NP_001157109.1 5 - 6 1410 74
GenBank mRNA AK133750.1 - 5 - 6 1375 74
AK143449.1 - 5 - 6 1178 74

View Full Product Details