The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109973.2 | NP_001103443.1 | 4 - 5 | 880 | 78 |
NM_001276390.1 | NP_001263319.1 | 3 - 4 | 762 | 78 | |
NM_001276391.1 | NP_001263320.1 | 5 - 6 | 652 | 78 | |
NM_010783.3 | NP_034913.2 | 4 - 5 | 548 | 78 | |
GenBank mRNA | BC006018.1 | - | 3 - 4 | 515 | 78 |
BC010259.1 | - | 4 - 5 | 720 | 78 | |
BC083058.1 | - | 4 - 5 | 653 | 78 | |
BC085233.1 | - | 4 - 5 | 880 | 78 | |
CX564914.1 | - | 4 - 5 | 619 | 78 | |
U57820.1 | - | 3 - 4 | 520 | 78 |