The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138649.1 | NP_619590.1 | 7 - 8 | 1420 | 126 |
GenBank mRNA | AB069667.1 | - | 7 - 8 | 1420 | 126 |
AK135098.1 | - | 5 - 6 | 1214 | 126 | |
AK144560.1 | - | 7 - 8 | 1597 | 126 |