The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130443.1 | NP_001123915.1 | 1 - 2 | 165 | 132 |
NM_001130444.1 | NP_001123916.1 | 1 - 2 | 165 | 132 | |
NM_008284.2 | NP_032310.2 | 1 - 2 | 165 | 132 | |
GenBank mRNA | BC050837.1 | - | 1 - 2 | 155 | 132 |
BC061885.1 | - | - | 121 | 132 | |
BQ963436.1 | - | 1 - 2 | 154 | 132 | |
CK788792.1 | - | 1 - 2 | 165 | 132 |