The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008284.2 | NP_032310.2 | 5 - 6 | 787 | 85 |
GenBank mRNA | BC050837.1 | - | 4 - 5 | 1440 | 85 |
BC061885.1 | - | - | 743 | 85 |