The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016891.3 | NP_058587.1 | 12 - 13 | 1575 | 76 |
GenBank mRNA | AB021743.1 | - | 12 - 13 | 1523 | 76 |
AK054239.1 | - | 12 - 13 | 1535 | 76 | |
AK078135.1 | - | 12 - 13 | 1554 | 76 | |
AK088776.1 | - | 12 - 13 | 1547 | 76 | |
AK180835.1 | - | 3 - 4 | 266 | 76 | |
BC006606.1 | - | 12 - 13 | 1575 | 76 | |
BC052678.1 | - | 12 - 13 | 1571 | 76 |