Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc94
Entrez Gene ID
72886
Gene Name
coiled-coil domain containing 94
Gene Aliases
2900016D05Rik, AI413813
Chromosome Location
Chr.17: 55959187 - 55968285 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028381.3 NP_082657.1 2 - 3 178 87
GenBank mRNA AK013533.1 - 2 - 3 192 87
AK168621.1 - 2 - 3 267 87
BC031400.1 - 2 - 3 178 87

View Full Product Details