Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc94
Entrez Gene ID
72886
Gene Name
coiled-coil domain containing 94
Gene Aliases
2900016D05Rik, AI413813
Chromosome Location
Chr.17: 55959187 - 55968285 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028381.3 NP_082657.1 4 - 5 460 67
GenBank mRNA AK168621.1 - 4 - 5 549 67
BC031400.1 - 4 - 5 460 67

View Full Product Details