The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053247.4 | NP_444477.2 | 5 - 6 | 862 | 87 |
GenBank mRNA | AJ311501.1 | - | 5 - 6 | 1017 | 87 |
AK004726.2 | - | 5 - 6 | 862 | 87 | |
AK160857.1 | - | 4 - 5 | 600 | 87 | |
BC038653.1 | - | 5 - 6 | 968 | 87 | |
BC038892.1 | - | 5 - 6 | 968 | 87 |