Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp5s
Entrez Gene ID
68055
Gene Name
ATP synthase, H+ transporting, mitochondrial F0 complex, subunit S
Gene Aliases
1110015E18Rik, ATPW
Chromosome Location
Chr.12: 69724930 - 69744665 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026536.1 NP_080812.1 2 - 3 157 105
GenBank mRNA AK003725.1 - 2 - 3 157 105
AK009141.1 - 3 - 4 269 105
BC027442.1 - 2 - 3 139 105

View Full Product Details