Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubxn4
Entrez Gene ID
67812
Gene Name
UBX domain protein 4
Gene Aliases
1300013G12Rik, Ubxd2, mKIAA0242
Chromosome Location
Chr.1: 128243898 - 128279378 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026390.2 NP_080666.2 4 - 5 622 83
GenBank mRNA AK011233.1 - 4 - 5 519 83
AK012824.1 - 4 - 5 516 83
AK019371.1 - 4 - 5 515 83
AK029431.1 - 4 - 5 668 83
AK032206.1 - 4 - 5 501 83
AK075716.1 - 4 - 5 357 83
AK145551.1 - 1 - 2 48 83
AK147795.1 - 4 - 5 495 83
AK148333.1 - 4 - 5 508 83
BC012271.1 - 4 - 5 634 83
BC019795.1 - 4 - 5 641 83
BC030190.1 - 4 - 5 631 83

View Full Product Details