Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Heatr6
- Entrez Gene ID
-
217026
- Gene Name
- HEAT repeat containing 6
- Gene Aliases
-
2700008B19Rik, BB135312, BB249351
- Chromosome Location
-
Chr.11: 83753397 - 83783754 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK169901.1 |
- |
1 - 2 |
252 |
62 |
View Full Product Details