Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amacr
Entrez Gene ID
17117
Gene Name
alpha-methylacyl-CoA racemase
Gene Aliases
Macr1
Chromosome Location
Chr.15: 10981756 - 10996624 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008537.4 NP_032563.2 1 - 2 297 62
GenBank mRNA AK002401.1 - 1 - 2 265 62
AK048249.1 - 1 - 2 298 62
AK160586.1 - 1 - 2 300 62
BC015825.1 - 1 - 2 225 62
U89906.1 - 1 - 2 205 62

View Full Product Details