Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc38
Entrez Gene ID
239570
Gene Name
tetratricopeptide repeat domain 38
Gene Aliases
9030019H09
Chromosome Location
Chr.15: 85832304 - 85858822 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033337.4 NP_001028509.2 10 - 11 997 84
GenBank mRNA AK133636.1 - 9 - 10 705 84
BC024550.1 - 10 - 11 947 84
BC030849.1 - 10 - 11 960 84
BC132621.1 - 10 - 11 982 84
BC132623.1 - 10 - 11 982 84

View Full Product Details