Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc38
Entrez Gene ID
239570
Gene Name
tetratricopeptide repeat domain 38
Gene Aliases
9030019H09
Chromosome Location
Chr.15: 85832304 - 85858822 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033337.4 NP_001028509.2 5 - 6 627 87
GenBank mRNA AK078848.1 - 5 - 6 588 87
BC024550.1 - 5 - 6 577 87
BC030849.1 - 5 - 6 590 87
BC132621.1 - 5 - 6 612 87
BC132623.1 - 5 - 6 612 87

View Full Product Details