Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mdga1
Entrez Gene ID
74762
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 1
Gene Aliases
1200011I03Rik, GPIM, Mamdc3
Chromosome Location
Chr.17: 29827956 - 29888429 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK004706.1 - 3 - 4 584 74
DQ788983.1 - - 3003 74

View Full Product Details