Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr.12: 81860028 - 82000924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018814.3 NP_061284.2 24 - 25 4930 105
GenBank mRNA AF096286.1 - 18 - 19 2760 105
AK129221.1 - 19 - 20 2810 105
AK154122.1 - 5 - 6 658 105
AK170320.1 - 5 - 6 658 105
BC057333.1 - 17 - 18 2673 105
BC066849.1 - 19 - 20 2590 105
BC098370.1 - 19 - 20 2520 105

View Full Product Details