Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- D2hgdh
- Entrez Gene ID
-
98314
- Gene Name
- D-2-hydroxyglutarate dehydrogenase
- Gene Aliases
-
AA408776, AA408778, AI325464
- Chromosome Location
-
Chr.1: 93824808 - 93852358 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK141691.1 |
- |
1 - 2 |
314 |
67 |
View Full Product Details