Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thsd7b
Entrez Gene ID
210417
Gene Name
thrombospondin, type I, domain containing 7B
Gene Aliases
1700074E13Rik, B930082A18, D130067I03Rik
Chromosome Location
Chr.1: 129273304 - 130219278 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172485.3 NP_766073.2 24 - 25 4711 75
GenBank mRNA BC063250.1 - 24 - 25 4541 75

View Full Product Details