Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsbn1
Entrez Gene ID
229675
Gene Name
rosbin, round spermatid basic protein 1
Gene Aliases
C230004D03Rik, E330004N01, Rosbin, Rsbn, Rsbp, mKIAA3002
Chromosome Location
Chr.3: 103914067 - 103966620 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172684.2 NP_766272.2 1 - 2 1030 91
GenBank mRNA AB101658.1 - 1 - 2 1032 91
AK122577.1 - 1 - 2 565 91
AK138603.1 - 1 - 2 574 91
BC069993.1 - 1 - 2 569 91
BC094667.1 - 1 - 2 561 91
BC139213.1 - 1 - 2 999 91
BC139215.1 - 1 - 2 999 91

View Full Product Details