The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178363.3 | NP_848140.3 | 14 - 15 | 6008 | 69 |
GenBank mRNA | AB033168.1 | - | 14 - 15 | 3681 | 69 |
AK028731.1 | - | 4 - 5 | 444 | 69 | |
AK044440.1 | - | 8 - 9 | 2325 | 69 | |
AK162250.1 | - | 6 - 7 | 718 | 69 | |
CA980613.1 | - | 2 - 3 | 231 | 69 |