The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178363.3 | NP_848140.3 | 1 - 2 | 1036 | 75 |
GenBank mRNA | AB033168.1 | - | 2 - 3 | 827 | 75 |
AK044440.1 | - | 1 - 2 | 1037 | 75 | |
AK087500.1 | - | 1 - 2 | 1007 | 75 |