Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h13
Entrez Gene ID
67302
Gene Name
zinc finger CCCH type containing 13
Gene Aliases
2600010B19Rik, 3110050K21Rik, 4930570G11Rik, C87618
Chromosome Location
Chr.14: 75284308 - 75344433 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026083.2 NP_080359.2 11 - 12 2394 61
GenBank mRNA AK078225.1 - 11 - 12 2385 61
BC038882.1 - 4 - 5 1204 61
BC094575.1 - 3 - 4 998 61
BC138264.1 - 11 - 12 2343 61

View Full Product Details