Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Disp1
Entrez Gene ID
68897
Gene Name
dispatched homolog 1 (Drosophila)
Gene Aliases
1190008H24Rik, DispA
Chromosome Location
Chr.1: 183086264 - 183221536 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026866.3 NP_081142.3 2 - 3 291 108
GenBank mRNA AK138276.1 - 2 - 3 248 108
BQ964652.1 - - 296 108

View Full Product Details