Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Homer2
Entrez Gene ID
26557
Gene Name
homer homolog 2 (Drosophila)
Gene Aliases
9330120H11Rik, AW539445, CPD, Vesl-2
Chromosome Location
Chr.7: 81600481 - 81706925 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164086.1 NP_001157558.1 2 - 3 321 94
NM_001164087.1 NP_001157559.1 2 - 3 304 94
NM_011983.2 NP_036113.1 2 - 3 321 94
GenBank mRNA AB017136.1 - 2 - 3 347 94
AF093259.1 - 2 - 3 226 94
AF093260.1 - 2 - 3 226 94
AJ012076.1 - 2 - 3 173 94
AK158673.1 - - 263 94
AK163423.1 - 2 - 3 322 94
DV657327.1 - 2 - 3 226 94

View Full Product Details