Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ndst3
Entrez Gene ID
83398
Gene Name
N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 3
Gene Aliases
4921531K01Rik, 4930511P15Rik, N-HSST 3, NDST-3
Chromosome Location
Chr.3: 123526166 - 123690852 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293682.1 NP_001280611.1 10 - 11 1438 103
NM_031186.3 NP_112463.2 11 - 12 2551 103
NR_121616.1 12 - 13 2655 103
GenBank mRNA AK014990.1 - 9 - 10 1591 103
AK015768.1 - 10 - 11 1438 103
AK141945.1 - 12 - 13 2656 103
BC079622.1 - 11 - 12 2422 103

View Full Product Details