The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135151.1 | NP_001128623.1 | 4 - 5 | 713 | 100 |
NM_001135152.1 | NP_001128624.1 | 4 - 5 | 679 | 100 | |
GenBank mRNA | AK090193.1 | - | 4 - 5 | 678 | 100 |
BC021530.1 | - | 4 - 5 | 716 | 100 |