The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013556.2 | NP_038584.2 | 8 - 9 | 752 | 95 |
GenBank mRNA | AK002286.1 | - | 8 - 9 | 697 | 95 |
AK088114.1 | - | 8 - 9 | 642 | 95 | |
BC004686.1 | - | 8 - 9 | 667 | 95 | |
BC083145.1 | - | 8 - 9 | 638 | 95 |