The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033272.1 | NP_001028444.1 | 12 - 13 | 3832 | 97 |
GenBank mRNA | AK151394.1 | - | 9 - 10 | 1714 | 97 |
AK158193.1 | - | 4 - 5 | 1366 | 97 | |
AK220313.1 | - | 9 - 10 | 1554 | 97 | |
BC035942.1 | - | 10 - 11 | 1850 | 97 | |
BC040408.1 | - | 2 - 3 | 439 | 97 | |
BC040810.1 | - | 2 - 3 | 439 | 97 | |
BC138455.1 | - | 12 - 13 | 3789 | 97 | |
BC145285.1 | - | 12 - 13 | 3789 | 97 |