Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dmtf1
Entrez Gene ID
23857
Gene Name
cyclin D binding myb-like transcription factor 1
Gene Aliases
Dimp, Dmp1
Chromosome Location
Chr.5: 9118868 - 9161776 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110327.1 NP_001103797.1 8 - 9 972 66
NM_011806.3 NP_035936.3 8 - 9 972 66
GenBank mRNA AK029147.1 - 8 - 9 914 66
AK031887.1 - 7 - 8 790 66
AK039563.1 - 8 - 9 940 66
AK078015.1 - 8 - 9 875 66
AK160595.1 - 8 - 9 945 66
BC045141.1 - 7 - 8 792 66
BC060109.1 - 8 - 9 894 66
U70017.1 - 8 - 9 927 66

View Full Product Details