The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252534.1 | NP_001239463.1 | 2 - 3 | 631 | 77 |
NM_001252536.1 | NP_001239465.1 | 3 - 4 | 355 | 77 | |
NM_001252537.1 | NP_001239466.1 | 2 - 3 | 631 | 77 | |
NM_001252538.1 | NP_001239467.1 | 2 - 3 | 321 | 77 | |
NM_028412.2 | NP_082688.1 | 2 - 3 | 273 | 77 | |
GenBank mRNA | AK089986.1 | - | 2 - 3 | 634 | 77 |
AK137961.1 | - | 2 - 3 | 237 | 77 | |
AK159103.1 | - | 2 - 3 | 609 | 77 | |
AK165687.1 | - | 3 - 4 | 309 | 77 | |
BC018483.1 | - | 2 - 3 | 237 | 77 | |
BC072624.1 | - | - | 476 | 77 | |
BY251304.1 | - | 2 - 3 | 322 | 77 |