The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289530.1 | NP_001276459.1 | 1 - 2 | 461 | 60 |
NM_052977.5 | NP_443209.2 | 1 - 2 | 461 | 60 | |
NR_110345.1 | 1 - 2 | 461 | 60 | ||
GenBank mRNA | AF232942.1 | - | 1 - 2 | 78 | 60 |
AF270495.1 | - | 1 - 2 | 178 | 60 | |
AK033916.1 | - | 1 - 2 | 400 | 60 | |
AK045538.1 | - | 1 - 2 | 400 | 60 | |
AK046083.1 | - | 1 - 2 | 405 | 60 | |
AK134964.1 | - | 1 - 2 | 463 | 60 | |
AK139079.1 | - | 1 - 2 | 419 | 60 | |
AK139273.1 | - | 1 - 2 | 419 | 60 | |
BC052426.1 | - | 1 - 2 | 386 | 60 | |
BC059822.1 | - | 1 - 2 | 414 | 60 |