Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Elf5
Entrez Gene ID
13711
Gene Name
E74-like factor 5
Gene Aliases
ESE-2, ESE-5, ESE-5.
Chromosome Location
Chr.2: 103411693 - 103450989 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145813.1 NP_001139285.1 6 - 7 694 67
NM_010125.3 NP_034255.2 6 - 7 785 67
GenBank mRNA AK132484.1 - 6 - 7 1105 67
AK145214.1 - 6 - 7 792 67
BC012424.1 - 6 - 7 727 67
BC021629.1 - 6 - 7 738 67

View Full Product Details