Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ighg3
Entrez Gene ID
380795
Gene Name
Immunoglobulin heavy constant gamma 3
Gene Aliases
IgG3
Chromosome Location
Chr.12: 113357442 - 113361232 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK037822.1 - - 676 84
AY571287.1 - - 720 84
BC055910.1 - - 747 84
D14625.1 - - 742 84
L36938.1 - 1 - 2 278 84

View Full Product Details