Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmd5
Entrez Gene ID
66998
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 5
Gene Aliases
1500032A03Rik, AW475925, S5b, W91691, mKIAA0072
Chromosome Location
Chr.2: 34852089 - 34870962 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080554.2 NP_542121.2 2 - 3 360 132
GenBank mRNA AK005335.1 - 2 - 3 338 132
AK011026.1 - 2 - 3 337 132
AK029067.1 - 2 - 3 331 132
AK032992.1 - 2 - 3 338 132
AK078419.1 - 2 - 3 338 132
AK145587.1 - 2 - 3 337 132
AK148284.1 - - 361 132
AK220321.1 - 2 - 3 301 132
BC019112.1 - 2 - 3 371 132

View Full Product Details