Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stt3b
Entrez Gene ID
68292
Gene Name
STT3, subunit of the oligosaccharyltransferase complex, homolog B (S. cerevisiae)
Gene Aliases
1300006C19Rik, Simp
Chromosome Location
Chr.9: 115242581 - 115310421 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024222.2 NP_077184.2 2 - 3 779 76
GenBank mRNA AK012153.1 - 2 - 3 449 76
AK018758.2 - 2 - 3 489 76
AK145674.1 - 2 - 3 420 76
AK152899.1 - 2 - 3 827 76
AK154979.1 - 2 - 3 468 76
AK170079.1 - 2 - 3 491 76
AX799084.1 - 2 - 3 490 76
BC052433.1 - 2 - 3 779 76

View Full Product Details