Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B4galt4
Entrez Gene ID
56375
Gene Name
UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 4
Gene Aliases
9130402O08Rik, B4galt-IV, b4Gal-T4, beta4Gal-T4, beta4GalT-IV
Chromosome Location
Chr.16: 38742103 - 38769054 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285793.1 NP_001272722.1 6 - 7 1222 58
NM_019804.4 NP_062778.2 6 - 7 1155 58
GenBank mRNA AF142672.1 - 6 - 7 1132 58
AF158746.1 - 6 - 7 1134 58
AK018658.1 - 6 - 7 1057 58
AK041692.1 - 6 - 7 1147 58
AK050005.1 - 6 - 7 1151 58
AK052013.1 - 6 - 7 1149 58
AK053603.1 - 6 - 7 1216 58
AK085368.1 - 6 - 7 1141 58
AK136042.1 - 2 - 3 327 58
BC013492.1 - 6 - 7 1118 58
BC031115.1 - 6 - 7 1207 58

View Full Product Details