Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abcc1
Entrez Gene ID
17250
Gene Name
ATP-binding cassette, sub-family C (CFTR/MRP), member 1
Gene Aliases
Abcc1a, Abcc1b, MRP, Mdrap, Mrp1
Chromosome Location
Chr.16: 14361521 - 14474879 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008576.3 NP_032602.1 3 - 4 386 75
GenBank mRNA AF022908.1 - 3 - 4 348 75
AK029876.1 - 3 - 4 387 75
AK150015.1 - 3 - 4 367 75
AK152781.1 - 3 - 4 367 75
BC090617.1 - 3 - 4 357 75

View Full Product Details