The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277875.1 | NP_001264804.1 | 4 - 5 | 698 | 66 |
NM_001277876.1 | NP_001264805.1 | 4 - 5 | 698 | 66 | |
NM_009416.4 | NP_033442.2 | 4 - 5 | 698 | 66 | |
GenBank mRNA | AK003186.1 | - | 4 - 5 | 609 | 66 |
BC014809.1 | - | 4 - 5 | 582 | 66 | |
BC024358.1 | - | 4 - 5 | 596 | 66 | |
CV560790.1 | - | 4 - 5 | 473 | 66 | |
DV049735.1 | - | 4 - 5 | 627 | 66 | |
M81086.1 | - | 4 - 5 | 532 | 66 | |
M87635.1 | - | 4 - 5 | 591 | 66 | |
X12650.1 | - | 4 - 5 | 593 | 66 | |
X58381.1 | - | 4 - 5 | 574 | 66 |