Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdha
Entrez Gene ID
66945
Gene Name
succinate dehydrogenase complex, subunit A, flavoprotein (Fp)
Gene Aliases
1500032O14Rik, 2310034D06Rik, 4921513A11, C81073, FP, SDH2, SDHF
Chromosome Location
Chr.13: 74322255 - 74350240 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023281.1 NP_075770.1 11 - 12 1585 92
GenBank mRNA AF095938.1 - 9 - 10 1334 92
AK004362.1 - 1 - 2 136 92
AK010561.1 - 11 - 12 1574 92
AK029520.1 - 11 - 12 1585 92
AK034928.1 - 11 - 12 1586 92
AK049590.1 - 11 - 12 1572 92
AK050475.1 - 11 - 12 1572 92
AK075990.1 - 11 - 12 1569 92
AK145923.1 - 11 - 12 1583 92
AK147286.1 - 11 - 12 1572 92
AK147624.1 - 11 - 12 1572 92
AK151333.1 - 3 - 4 213 92
AK153085.1 - 11 - 12 1592 92
AK162148.1 - 11 - 12 1568 92
AK169254.1 - 11 - 12 1568 92
BC011301.1 - 11 - 12 1545 92
BC031849.1 - 11 - 12 1593 92

View Full Product Details