The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009951.1 | NP_001009951.1 | 10 - 11 | 1189 | 124 |
GenBank mRNA | AK220415.1 | - | 9 - 10 | 1145 | 124 |
BC088983.1 | - | 10 - 11 | 1189 | 124 |