Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cep44
Entrez Gene ID
382010
Gene Name
centrosomal protein 44
Gene Aliases
4933440G23Rik, mKIAA1712
Chromosome Location
Chr.8: 56531516 - 56550777 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009951.1 NP_001009951.1 9 - 10 1144 115
GenBank mRNA AK220415.1 - 8 - 9 1100 115
BC088983.1 - 9 - 10 1144 115

View Full Product Details