Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Invs
Entrez Gene ID
16348
Gene Name
inversin
Gene Aliases
Nphp2, inv
Chromosome Location
Chr.4: 48279760 - 48431953 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281977.1 NP_001268906.1 13 - 14 2263 111
NM_001281978.1 NP_001268907.1 15 - 16 2541 111
NM_010569.4 NP_034699.3 13 - 14 2315 111
GenBank mRNA AJ010902.1 - 13 - 14 2256 111
AK041671.1 - 11 - 12 1975 111
AK144794.1 - 1 - 2 245 111
AK158766.1 - 15 - 16 2500 111
BC126929.1 - 13 - 14 2152 111

View Full Product Details