The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033962.1 | NP_001029134.1 | 7 - 8 | 2620 | 155 |
NM_011668.2 | NP_035798.2 | 9 - 10 | 2807 | 155 | |
NM_173010.3 | NP_766598.1 | 7 - 8 | 2622 | 155 | |
GenBank mRNA | AK029133.1 | - | 7 - 8 | 2623 | 155 |
AK220549.1 | - | 7 - 8 | 2520 | 155 | |
BC131658.1 | - | 7 - 8 | 2608 | 155 | |
BC131659.1 | - | 7 - 8 | 2608 | 155 | |
U82122.1 | - | 5 - 6 | 2057 | 155 | |
U96636.1 | - | 9 - 10 | 155 |